3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
A822485-25G
A822485-5G
A822485-10G
A822485-1G
103841-140EA
13
USD
103841-140
76845-376
76845-378
77281-504
77308-400
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine 98%
3-Bromo-2-chloro-5-(trifluoromethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₂BrClF₃N Boiling Pt: 210.5 °C (760 mm Hg) Melting Pt: 28…32 °C Flash Pt: 98.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09878432 CAS Number: 71701-92-3 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...