3-Bromo-2-chloro-4-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-chloro-4-picoline

A112760-100G A112760-10G A112760-25G A112760-1G A112760-5G
103798-096EA 6 USD
103798-096 103823-934 76970-942 76970-944 76970-946 77310-754 77310-756
3-Bromo-2-chloro-4-methylpyridine 98%
3-Bromo-2-chloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Melting Pt: 64…69 °C
Storage Temperature: Ambient
MDL Number: MFCD04112511
CAS Number: 55404-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR