3-Bromo-2-chloro-4-methoxypyridine 97%

Supplier: Ambeed
Warning

A139648-100MG A139648-250MG A139648-1G
103802-754EA 79 USD
103802-754 77010-308 77010-310 77299-080
3-Bromo-2-chloro-4-methoxypyridine 97%
3-Bromo-2-chloro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.47 g/mol
Boiling Pt: 261.2±35.0 °C (760 mm Hg)
Melting Pt: 113…117 °C
Flash Pt: 111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD14705097
CAS Number: 144584-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR