3-Bromo-2,6-dimethylpyridine 98%
Supplier: Ambeed
|
Danger
|
A172705-1G
A172705-25G
A172705-5G
A172705-500G
103806-738EA
10
USD
103806-738
103845-032
77082-238
77082-240
77082-242
77191-480
3-Bromo-2,6-dimethylpyridine 98%
3-Bromo-2,6-dimethylpyridine
Precaución: for Research Use Only
Formula:
C₇H₈BrN Boiling Pt: 194.5 °C (760 mm Hg) Flash Pt: 71.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00235164 CAS Number: 3430-31-7 UN: 1760 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...