3-Bromo-2,6-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A453579-100G A453579-100MG A453579-10G A453579-1G A453579-25G A453579-5G A453579-250MG
103827-812EA 9 USD
103827-812 103838-882 76818-376 76818-378 76818-380 76842-712 76842-714 76842-716 77673-134
3-Bromo-2,6-difluorobenzoic acid 97%
3-Bromo-2,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MW: 237.0000 g/mol
Boiling Pt: 279.2 °C (760 mm Hg)
Melting Pt: 137…139 °C
Flash Pt: 123 °C
Storage Temperature: Ambient
MDL Number: MFCD09261248
CAS Number: 28314-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR