3-Bromo-2,4-dimethylpyridine 96%

Supplier: Ambeed
Warning

A215036-100MG A215036-250MG
103818-450EA 189 USD
103818-450 76785-796 77667-800
3-Bromo-2,4-dimethylpyridine 96%
3-Bromo-2,4-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD09842453
CAS Number: 27063-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR