3-Bromo-2-(chloromethyl)pyridine 98%

Supplier: Ambeed
Danger

A454033-1G A454033-250MG
103827-820EA 20 USD
103827-820 103838-890 76842-728 77016-782
3-Bromo-2-(chloromethyl)pyridine 98%
3-Bromo-2-(chloromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 237.949 °C (760 mm Hg)
Flash Pt: 97.709 °C
Storage Temperature: Freezer
MDL Number: MFCD13189055
CAS Number: 122851-69-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR