3-Bromo-1-methylpyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A281878-1G A281878-5G A281878-100MG A281878-10G A281878-250MG A281878-25G
103813-576EA 108 USD
103813-576 103853-528 76837-228 76837-230 76988-494 76988-496 76988-498 76988-500
3-Bromo-1-methylpyridin-2(1H)-one 95%
3-Bromo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD11226865
CAS Number: 81971-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR