3-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

A110749-100MG A110749-1G A110749-5G
103797-718EA 96 USD
103797-718 103823-528 77008-898 77284-030 77284-032
3-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine 95%
3-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11878034
CAS Number: 116855-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR