3-Bromo-1H-pyrrolo[3,2-b]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-4-azaindole

A116011-100MG A116011-10G A116011-250MG A116011-1G
103798-658EA 34 USD
103798-658 103824-522 77026-102 77026-104 77026-106 77311-344
3-Bromo-1H-pyrrolo[3,2-b]pyridine 98%
3-Bromo-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697525
CAS Number: 23688-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR