3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%

Supplier: Ambeed
Warning

A517427-250MG A517427-5G A517427-1G
103828-472EA 29 USD
103828-472 103839-576 76790-058 76843-570 77307-690
3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%
3-Bromo-1H-pyrazolo[4,3-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
Boiling Pt: 366.9 °C (760 mm Hg)
Flash Pt: 175.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11518974
CAS Number: 633328-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR