3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 96%

Supplier: Ambeed
Warning

A234857-10G A234857-25G A234857-5G A234857-1G
103810-984EA 5 USD
103810-984 103850-744 77031-156 77031-158 77089-744 77304-004
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine 96%
3-Bromo-1H-pyrazolo[3,4-d]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C5H4BrN5
Storage Temperature: Refrigerator
MDL Number: MFCD12196922
CAS Number: 83255-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR