3-Bromo-1H-pyrazolo[3,4-b]pyridine 97%
Supplier: Ambeed
|
Warning
|
A104020-10G
A104020-25G
A104020-1G
A104020-5G
103796-640EA
14
USD
103796-640
103822-352
77233-166
77233-168
77309-212
77309-214
3-Bromo-1H-pyrazolo[3,4-b]pyridine 97%
3-Bromo-1H-pyrazolo[3,4-b]pyridine
Precaución: for Research Use Only
Formula:
C₆H₄BrN₃ MW: 198.0200 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD08669476 CAS Number: 68618-36-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...