3-Bromo-1H-pyrazole 98%

Supplier: Ambeed
Warning

A120614-100G A120614-10G A120614-250MG A120614-25G A120614-1G A120614-5G
103799-968EA 6 USD
103799-968 77108-562 77108-564 77108-566 77108-568 77312-074 77312-076
3-Bromo-1H-pyrazole 98%
3-Bromo-1H-pyrazole

Precaución: for Research Use Only

Formula: C3H3BrN2
MW: 146.97 g/mol
Melting Pt: 60…65 °C
Flash Pt: 113.5 °C
Storage Temperature: Freezer
MDL Number: MFCD01646049
CAS Number: 14521-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR