3-Bromo-1H-indazole-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A219214-100MG A219214-250MG A219214-5G
103818-772EA 79 USD
103818-772 76785-916 76785-918 76785-920
3-Bromo-1H-indazole-7-carboxylic acid 95%
3-Bromo-1H-indazole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.04 g/mol
Boiling Pt: 493.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11977526
CAS Number: 1040101-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR