3-Bromo-1H-indazole-7-carbonitrile 95%

Supplier: Ambeed
Warning

A366631-250MG A366631-1G
103826-406EA 108 USD
103826-406 103837-422 76788-622 77306-780
3-Bromo-1H-indazole-7-carbonitrile 95%
3-Bromo-1H-indazole-7-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848409
CAS Number: 945762-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR