3-Bromo-1H-indazole 97%

Supplier: Ambeed
Danger

A153191-10G A153191-250MG A153191-1G A153191-25G A153191-5G
103804-470EA 14 USD
103804-470 103842-640 77273-032 77273-034 77299-998 77300-000 77300-002
3-Bromo-1H-indazole 97%
3-Bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
Boiling Pt: 179.2 °C (760 mm Hg)
Melting Pt: 143…147 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159926
CAS Number: 40598-94-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR