3-Bromo-1,7-naphthyridin-8(7H)-one 97%
Supplier: Ambeed
|
Warning
|
A378639-1G
A378639-5G
A378639-100MG
A378639-10G
A378639-250MG
103826-630EA
66
USD
103826-630
103837-664
76840-974
76840-976
77240-342
77240-344
77240-346
3-Bromo-1,7-naphthyridin-8(7H)-one 97%
3-Bromo-1,7-naphthyridin-8(7H)-one
Precaución: for Research Use Only
Formula:
C₈H₅BrN₂O Boiling Pt: 468.9 °C (760 mm Hg) Flash Pt: 237.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD22548037 CAS Number: 1375301-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...