3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95%

Supplier: Ambeed
Warning

A458055-250MG A458055-1G A458055-5G
103827-882EA 57 USD
103827-882 103838-958 76979-526 77307-406 77307-408
3-Bromo[1,2,4]triazolo[4,3-a]pyridine 95%
3-Bromo[1,2,4]triazolo[4,3-a]pyridine

Precaución: for Research Use Only

Formula: C6H4BrN3
MW: 198.0200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11111699
CAS Number: 4922-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR