3-Carboxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

A154506-10G A154506-500G A154506-5G A154506-25G A154506-100G
103842-840EA 5 USD
103842-840 76768-276 76768-278 77300-064 77318-570 77592-896
3-Carboxyphenylboronic acid 98%
3-Carboxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BO₄
MW: 165.94 g/mol
Boiling Pt: 434.5 °C (760 mm Hg)
Melting Pt: 243…247 °C
Flash Pt: 216.585 °C
Storage Temperature: Ambient
MDL Number: MFCD00036833
CAS Number: 25487-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR