3-Carboxy-5-nitrophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Carboxy-5-nitrobenzeneboronic acid, 3-Borono-5-nitrobenzoic acid
A209915-1G
A209915-5G
A209915-250MG
103817-968EA
6
USD
103817-968
103848-654
77087-018
77087-020
77139-576
3-Carboxy-5-nitrophenylboronic acid 97%
3-Carboxy-5-nitrophenylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₆BNO₆ MW: 210.94 g/mol Melting Pt: 248…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00757433 CAS Number: 101084-81-5 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...