3-Boc-aminopyrrolidine 98%

Supplier: Ambeed
Warning

Synonyms: 3-(BOC-amino)pyrrolidine, 2-Methyl-2-propanyl 3-pyrrolidinylcarbamate, 1,1-Dimethylethyl3-pyrrolidinylcarbamate, 3-Boc-Aminopyrrolidine

A100593-100G A100593-1G A100593-5G A100593-25G
103795-530EA 9 USD
103795-530 103821-218 77257-212 77296-772 77296-774 77548-886
3-Boc-aminopyrrolidine 98%
3-Boc-aminopyrrolidine

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂
Melting Pt: 63…68 °C
Flash Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00059040
CAS Number: 99724-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR