tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 3-Boc-3,8-diazabicyclo[3.2.1]octane, 3,8-Diazabicyclo[3.2.1]octane, N-BOC protected

A172920-100MG A172920-10G A172920-250MG A172920-1G A172920-5G A172920-100G
103806-764EA 18 USD
103806-764 103845-056 77029-152 77029-154 77029-156 77082-274 77082-276 77600-402
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate 97%
tert-Butyl 3,8-diazabicyclo[3.2.1]octane-3-carboxylate

Precaución: for Research Use Only

Formula: C11H20N2O2
MW: 212.29 g/mol
Flash Pt: 132.451 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17016700
CAS Number: 201162-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR