(1R,5S)-3-Benzyl-3-azabicyclo[3.2.1]octan-8-one 97%

Supplier: Ambeed
Warning

Synonyms: 3-Benzyl-3-azabicyclo[3.2.1]octan-8-one

A190701-1G A190701-25G A190701-5G A190701-10G A190701-250MG
103816-320EA 8 USD
103816-320 103846-922 77084-734 77084-736 77084-738 77274-410 77274-412
(1R,5S)-3-Benzyl-3-azabicyclo[3.2.1]octan-8-one 97%
(1R,5S)-3-Benzyl-3-azabicyclo[3.2.1]octan-8-one

Precaución: for Research Use Only

Formula: C₁₄H₁₇NO
Boiling Pt: 342.9 °C (760 mm Hg)
Flash Pt: 151 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08275096
CAS Number: 83507-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR