3-Benzyl-3,9-diazaspiro[5.5]undecane 95%

Supplier: Ambeed
Warning

A406172-1G A406172-5G A406172-100MG A406172-250MG
103827-132EA 54 USD
103827-132 103838-170 76841-662 76841-664 77278-824 77278-826
3-Benzyl-3,9-diazaspiro[5.5]undecane 95%
3-Benzyl-3,9-diazaspiro[5.5]undecane

Precaución: for Research Use Only

Formula: C₁₆H₂₄N₂
MW: 244.375 g/mol
Boiling Pt: 357.7±42.0 °C (760 mm Hg)
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD11035940
CAS Number: 189333-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR