3-Benzoylpropionic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid

A111625-100G A111625-10G A111625-25G A111625-500G A111625-5G
103823-692EA 8 USD
103823-692 77270-956 77270-958 77270-960 77270-962 77284-150
3-Benzoylpropionic acid 95%
3-Benzoylpropionic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 359.7 °C (760 mm Hg)
Melting Pt: 118 °C
Flash Pt: 185.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002792
CAS Number: 2051-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR