(±)-3-Aminoquinuclidine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride

A373183-1G A373183-25G A373183-5G A373183-100G A373183-10G A373183-250MG
103826-528EA 11 USD
103826-528 103837-546 76840-806 76840-808 76840-810 77278-450 77278-452 77278-454
(±)-3-Aminoquinuclidine dihydrochloride 97%
(±)-3-Aminoquinuclidine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
Melting Pt: 321…323 °C
Storage Temperature: Ambient
MDL Number: MFCD00137395
CAS Number: 6530-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR