3-Aminoquinolin-4-ol 97%

Supplier: Ambeed
Warning

A751688-250MG A751688-1G
103840-854EA 594 USD
103840-854 76652-382 77308-254
3-Aminoquinolin-4-ol 97%
3-Aminoquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD09907852
CAS Number: 99512-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR