3-Aminopyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-1,2-dihydro-2-oxopyridine, 1,2-Dihydro-2-oxopyridin-3-amine

A286539-100G A286539-10G A286539-1G A286539-25G A286539-5G A286539-500G
103813-718EA 9 USD
103813-718 103834-854 76816-478 76816-480 76837-390 76837-392 76837-394 77883-777
3-Aminopyridin-2(1H)-one 98%
3-Aminopyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₆N₂O
Boiling Pt: 358.7 °C (760 mm Hg)
Melting Pt: 118…130 °C
Flash Pt: 170.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02752887
CAS Number: 33630-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR