3-Aminopyridazine 98%
Supplier: Ambeed
|
Warning
|
A123120-100G
A123120-10G
A123120-25G
A123120-1G
A123120-5G
103800-344EA
9
USD
103800-344
77108-680
77108-682
77108-684
77297-926
77312-464
3-Aminopyridazine 98%
3-Aminopyridazine
Precaución: for Research Use Only
Formula:
C4H5N3 Melting Pt: 168…172 °C Flash Pt: 177.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD01529869 CAS Number: 5469-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...