3-Aminopyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A240931-1G A240931-250MG
103811-378EA 94 USD
103811-378 103851-184 77090-344 77672-150
3-Aminopyrazolo[1,5-a]pyrimidine 97%
3-Aminopyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C6H6N4
MW: 134.1400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00501923
CAS Number: 232600-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR