2-Amino-3-hydroxypyrazine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Aminopyrazin-2(1H)-one

A148001-250MG A148001-25G A148001-1G
103803-862EA 29 USD
103803-862 103841-978 77247-160 77299-626 77317-408
2-Amino-3-hydroxypyrazine 97%
2-Amino-3-hydroxypyrazine

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MW: 111.1 g/mol
Boiling Pt: 538.9 °C (760 mm Hg)
Melting Pt: 300…301 °C
Storage Temperature: Ambient
MDL Number: MFCD01646052
CAS Number: 43029-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR