3-Aminophenyl dimethylcarbamate 97%

Supplier: Ambeed
Warning

A119757-1G A119757-250MG
103799-830EA 109 USD
103799-830 77297-718 77747-872
3-Aminophenyl dimethylcarbamate 97%
3-Aminophenyl dimethylcarbamate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
Boiling Pt: 313 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00052699
CAS Number: 19962-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR