3-Amino-2-naphthol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-2-hydroxynaphthalene, 3-Aminonaphthalen-2-ol
A175112-100MG
A175112-250MG
A175112-1G
A175112-5G
103807-002EA
41
USD
103807-002
103845-306
76799-506
76799-508
77082-630
77082-632
3-Amino-2-naphthol 97%
3-Amino-2-naphthol
Precaución: for Research Use Only
Formula:
C₁₀H₉NO MW: 159.19 g/mol Boiling Pt: 343.5 °C (760 mm Hg) Melting Pt: 232…238 °C Flash Pt: 161.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004113 CAS Number: 5417-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...