3-Aminoisonicotinamide 98%

Supplier: Ambeed
Warning

A136514-100MG A136514-10G A136514-250MG A136514-1G A136514-25G A136514-5G
103802-310EA 16 USD
103802-310 77272-252 77272-254 77272-256 77315-206 77315-208 77315-210
3-Aminoisonicotinamide 98%
3-Aminoisonicotinamide

Precaución: for Research Use Only

Formula: C₆H₇N₃O
Boiling Pt: 369.2 °C (760 mm Hg)
Melting Pt: 149…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08235178
CAS Number: 64188-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR