3-Aminocyclobutanone hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 3-aminocyclobutan-1-one hydrochloride

A303919-1G A303919-5G A303919-10G A303919-250MG A303919-25G A303919-100G
103814-402EA 13 USD
103814-402 103835-568 76838-256 76838-258 76977-762 76977-764 76977-766 77881-842
3-Aminocyclobutanone hydrochloride 97%
3-Aminocyclobutanone hydrochloride

Precaución: For research use only

Formula: C₄H₈ClNO
MW: 121.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923220
CAS Number: 1035374-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR