3-Aminobutan-1-ol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-butan-1-ol

A161784-100G A161784-10G A161784-500G A161784-1G A161784-25G A161784-5G
103805-538EA 6 USD
103805-538 103843-780 77273-332 77273-334 77273-336 77319-792 77319-794 77319-796
3-Aminobutan-1-ol 97%
3-Aminobutan-1-ol

Precaución: for Research Use Only

Formula: C₄H₁₁NO
Boiling Pt: 168.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03412694
CAS Number: 2867-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR