3-Amino-6-methylpyrazin-2(1H)-one 95%

Supplier: Ambeed
Warning

A240761-100MG A240761-250MG A240761-1G A240761-5G
103811-362EA 208 USD
103811-362 103851-166 77013-758 77013-760 77090-316 77090-318
3-Amino-6-methylpyrazin-2(1H)-one 95%
3-Amino-6-methylpyrazin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD19216347
CAS Number: 43029-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR