3-Amino-6-chloropyridine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A194190-1G A194190-5G A194190-100MG A194190-250MG
103816-630EA 12 USD
103816-630 103847-242 77085-160 77085-162 77274-572 77274-574
3-Amino-6-chloropyridine-2-carboxylic acid 98%
3-Amino-6-chloropyridine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C6H5ClN2O2
MW: 172.5691 g/mol
Boiling Pt: 413.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09264291
CAS Number: 866807-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR