3-Amino-6-chloropyridazine 98%

Supplier: Ambeed
Warning

A261847-100G A261847-10G A261847-500G A261847-1000G A261847-25G
103809-858EA 157 USD
103809-858 76816-018 76816-020 76816-022 77092-038 77092-040
3-Amino-6-chloropyridazine 98%
3-Amino-6-chloropyridazine

Precaución: for Research Use Only

Formula: C₄H₄ClN₃
Boiling Pt: 363.1 °C (760 mm Hg)
Melting Pt: 210 °C
Flash Pt: 173.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00051506
CAS Number: 5469-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR