3-Amino-6-bromopyridazine 97%

Supplier: Ambeed
Warning

A113305-10G A113305-250MG A113305-25G A113305-1G A113305-5G
103798-212EA 11 USD
103798-212 103824-052 76766-014 76766-016 76766-018 77310-866 77310-868
3-Amino-6-bromopyridazine 97%
3-Amino-6-bromopyridazine

Precaución: for Research Use Only

Formula: C4H4BrN3
Melting Pt: 194 °C
Flash Pt: 167.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01646092
CAS Number: 88497-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR