6-Bromo-1,2,4-triazin-3-amine 95%

Supplier: Ambeed
Warning

A160416-10G A160416-250MG A160416-1G A160416-5G A160416-100G
103805-376EA 6 USD
103805-376 103843-604 76985-032 76985-034 77319-536 77319-538 77611-558
6-Bromo-1,2,4-triazin-3-amine 95%
6-Bromo-1,2,4-triazin-3-amine

Precaución: for Research Use Only

Formula: C₃H₃BrN₄
Boiling Pt: 373.6 °C (760 mm Hg)
Flash Pt: 179.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16877209
CAS Number: 69249-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR