3-Amino-5-methylpyridazine 97%

Supplier: Ambeed
Warning

A338774-100MG A338774-250MG
103825-780EA 155 USD
103825-780 103836-754 76876-144 76876-146
3-Amino-5-methylpyridazine 97%
3-Amino-5-methylpyridazine

Precaución: for Research Use Only

Formula: C₅H₇N₃
MW: 109.1292 g/mol
Boiling Pt: 320.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD10698051
CAS Number: 144294-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR