3-Amino-5-methylbenzotrifluoride 95%

Supplier: Ambeed
Warning

A131823-250MG A131823-1G A131823-10G A131823-25G
103801-642EA 59 USD
103801-642 76688-654 77298-510 77876-941 77876-942
3-Amino-5-methylbenzotrifluoride 95%
3-Amino-5-methylbenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₈F₃N
Boiling Pt: 203.3 °C (760 mm Hg)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631580
CAS Number: 96100-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR