3-Amino-5-methoxybenzonitrile 95%
Supplier: Ambeed
|
Warning
|
A110591-100MG
A110591-10G
A110591-250MG
A110591-1G
A110591-5G
103797-688EA
75
USD
103797-688
103823-494
76821-768
76821-770
76821-772
77284-020
77284-022
3-Amino-5-methoxybenzonitrile 95%
3-Amino-5-methoxybenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₈N₂O MW: 148.16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18389726 CAS Number: 269411-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...