3-Amino-5-hydroxybenzoic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Amino-5-hydroxybenzoic acid, HCl
A279551-10G
A279551-1G
A279551-5G
103813-470EA
10
USD
103813-470
103853-416
77276-932
77305-334
77305-336
3-Amino-5-hydroxybenzoic acid hydrochloride 97%
3-Amino-5-hydroxybenzoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₈ClNO₃ MW: 189.6000 g/mol Boiling Pt: 453.4 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00043420 CAS Number: 14206-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...