3-Amino-5-hydroxybenzoic acid 97%

Supplier: Ambeed
Warning

A295566-1G A295566-5G A295566-100MG A295566-250MG
103814-064EA 59 USD
103814-064 103835-198 76837-856 76837-858 77277-248 77277-250
3-Amino-5-hydroxybenzoic acid 97%
3-Amino-5-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
Boiling Pt: 453.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD06797571
CAS Number: 76045-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR