3-Amino-5-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

A100506-250MG A100506-1G A100506-5G A100506-10G A100506-25G
103795-488EA 21 USD
103795-488 103821-182 76687-000 77282-504 77282-506 77623-810 77623-812
3-Amino-5-fluorobenzoic acid 97%
3-Amino-5-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Flash Pt: 168.833 °C
Storage Temperature: Freezer
MDL Number: MFCD07779526
CAS Number: 786616-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR