3-Amino-5-fluoro-6-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-6-methoxypyridin-3-amine

A350941-100MG A350941-250MG
103826-046EA 145 USD
103826-046 103837-042 76817-322 77594-048
3-Amino-5-fluoro-6-methoxypyridine 98%
3-Amino-5-fluoro-6-methoxypyridine

Precaución: for Research Use Only

Formula: C6H7FN2O
MW: 142.1300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07374960
CAS Number: 886372-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR