3-Amino-5-chlorophenol 97%
Supplier: Ambeed
|
Danger
|
A114197-250MG
A114197-5G
A114197-1G
103798-368EA
73
USD
103798-368
103824-208
76982-870
77297-296
77311-028
3-Amino-5-chlorophenol 97%
3-Amino-5-chlorophenol
Precaución: for Research Use Only
Formula:
C₆H₆ClNO Storage Temperature: Refrigerator |
MDL Number:
MFCD16999375 CAS Number: 883195-40-2 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...